Learn More
Keystone Cleanroom Products 1073B Tyvek™/HDPE Autoclave Stopper Bag Tubing
Autoclave Stopper Bag Continuous Form Tubing for Steam Sterilization is made of medical grade DuPont™ Tyvek™ and HDPE film materials and yields high seal strength for stoppers, seals and heavy equipment parts.
Brand:  Keystone Cleanroom Products 018SBT
 View more versions of this product
Product Code. 17871711
  • 22510.00 DKK / Pack of 500
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Designed for Steam Sterilization
  • Recommended for Use in Conjunction with a Secondary Tyvek™ Sterilization Pouch
  • Hydrophobic
  • Microbial Barrier
  • Resists Tearing and Punctures
  • Lint-Free - Category (Helmke Drum)
  • Manufactured and Packaged in a ISO 6 Cleanroom
  • Certificates of Conformance sent with shipment
  • Cut tubing to desired length and heat seal both ends closed using a validatable impulse, or continuous band sealer
Specifications
Specifications
Heat Seal
500 Pk.
500 ft.
18 in.
Single-use
500 ft. x 18 in.
152.4 m
45.7 cm