Learn More
Keystone Cleanroom Products 1073B Tyvek™/HDPE Autoclave Stopper Bag Tubing
Autoclave Stopper Bag Continuous Form Tubing for Steam Sterilization is made of medical grade DuPont™ Tyvek™ and HDPE film materials and yields high seal strength for stoppers, seals and heavy equipment parts.
13935.00 DKK - 22510.00 DKK
Specifications
Closure Type Heat Seal
Disposable Single-use
Quantity 500 Pk.
View More Specs
Products 2
Product Code Brand Width (English) Price Quantity & Availability  
Product Code Brand Width (English) Price Quantity & Availability  
17821711
View Documents
Keystone Cleanroom Products
14 in.
13935.00 DKK
Pack of 500
17871711
View Documents
Keystone Cleanroom Products
18 in.
22510.00 DKK
Pack of 500
Description
Description
  • Designed for Steam Sterilization
  • Recommended for Use in Conjunction with a Secondary Tyvek™ Sterilization Pouch
  • Hydrophobic
  • Microbial Barrier
  • Resists Tearing and Punctures
  • Lint-Free - Category (Helmke Drum)
  • Manufactured and Packaged in a ISO 6 Cleanroom
  • Certificates of Conformance sent with shipment
  • Cut tubing to desired length and heat seal both ends closed using a validatable impulse, or continuous band sealer
Specifications
Specifications
Heat Seal
Single-use
500 Pk.