Learn More
Keystone Cleanroom Products 1073B Tyvek™/HDPE Autoclave Stopper Bag Tubing
Autoclave Stopper Bag Continuous Form Tubing for Steam Sterilization is made of medical grade DuPont™ Tyvek™ and HDPE film materials and yields high seal strength for stoppers, seals and heavy equipment parts.
13935.00 DKK - 22510.00 DKK
Product Code | Brand | Width (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (English) | Price | Quantity & Availability | |||||
17821711
|
Keystone Cleanroom Products
|
14 in. |
13935.00 DKK
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17871711
|
Keystone Cleanroom Products
|
18 in. |
22510.00 DKK
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Designed for Steam Sterilization
- Recommended for Use in Conjunction with a Secondary Tyvek™ Sterilization Pouch
- Hydrophobic
- Microbial Barrier
- Resists Tearing and Punctures
- Lint-Free - Category (Helmke Drum)
- Manufactured and Packaged in a ISO 6 Cleanroom
- Certificates of Conformance sent with shipment
- Cut tubing to desired length and heat seal both ends closed using a validatable impulse, or continuous band sealer
Specifications
Heat Seal | |
Single-use | |
500 Pk. |