Learn More
Bochem™ 18/10 Stainless Steel Forceps, Sharp Tip
Assists with surgical procedures. Bochem™ 18/10 Stainless Steel Forceps, Sharp Tip are a very useful tool.
Brand: Bochem™ 1010
This item is not returnable.
View return policy
Description
- Stainless Steel
- Various lengths
Specifications
Sharp | |
105 mm | |
1 Pack |
Stainless Steel (18/10) | |
Forceps | |
Forceps |
Product Content Correction
Product Title
Bochem™ 18/10 Stainless Steel Forceps, Sharp Tip > Tip Style: Sharp; Length: 105mm; Quantity: 1/Pk
Spot an opportunity for improvement?Share a Content Correction