Learn More
2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%
CAS: 1159599-99-1 | C7H11NO5 | 189.17 g/mol
605.00 DKK - 7680.00 DKK
Chemical Identifiers
CAS 1159599-99-1
Molecular Formula C7H11NO5
Molecular Weight (g/mol) 189.17
MDL Number MFCD11976178
InChI Key KOUVDKDABFOPIG-UHFFFAOYSA-N
Synonym 2-oxa-6-azaspiro 3.3 heptane oxalate, 2-oxa-6-aza-spiro 3.3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3,3 heptane oxalic acid salt, 2-oxa-6-azaspiro 3.3 heptane xoxalate, 2-oxa-6-azaspiro 3.3 heptane oxalic acid, 2-oxa-6-azaspiro 3.3 heptane; oxalic acid, 2-oxa-6-azaspiro 3.3 heptane ethanedioate, 2-oxa-6-azaspiro 3.3 heptane oxalate 1:1, 2-oxa-6-azaspiro 3.3 heptane, ethanedioate 1:1, acmc-2099rj
PubChem CID 53308464
SMILES OC(=O)C(O)=O.C1NCC11COC1
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15470339
SDS View Documents
Thermo Scientific Alfa Aesar
H63578.MD
250 mg
605.00 DKK
250mg
15450339
SDS View Documents
Thermo Scientific Alfa Aesar
H63578.03
1 g
1870.00 DKK
1g
15460339
SDS View Documents
Thermo Scientific Alfa Aesar
H63578.06
5 g
7680.00 DKK
5g
Description
Description
2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Oxa-6-azaspiro[3.3]heptane oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications
Show More
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H301-H311
Toxic if swallowed.
Toxic in contact with skin.
P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
H302+H312
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
SDS
Documents
Documents
Product Certifications
RUO – Research Use Only
Product Content Correction
Product Title
2-Oxa-6-azaspiro[3.3]heptane oxalate, 97%