missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dimethyl-3-heptanol, 99%, Thermo Scientific™
1496.00 DKK - 6177.00 DKK
Chemical Identifiers
CAS | 1573-28-0 |
---|---|
Molecular Formula | C9H20O |
Molecular Weight (g/mol) | 144.26 |
MDL Number | MFCD00021828 |
InChI Key | HVPGGLNDHUWMLS-UHFFFAOYNA-N |
PubChem CID | 15297 |
IUPAC Name | 3,6-dimethylheptan-3-ol |
SMILES | CCC(C)(O)CCC(C)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
1573-28-0 | |
144.26 | |
HVPGGLNDHUWMLS-UHFFFAOYNA-N | |
3,6-dimethylheptan-3-ol |
C9H20O | |
MFCD00021828 | |
15297 | |
CCC(C)(O)CCC(C)C |
Specifications
1573-28-0 | |
C9H20O | |
3,6-dimethyl-3-heptanol, 3-heptanol, 3,6-dimethyl, acmc-20anbp, 3-heptanol,3,6-dimethyl | |
CCC(C)(O)CCC(C)C | |
144.26 | |
144.25 | |
3,6-Dimethyl-3-heptanol |
0.82 | |
MFCD00021828 | |
HVPGGLNDHUWMLS-UHFFFAOYNA-N | |
3,6-dimethylheptan-3-ol | |
15297 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures