Learn More
3'-Azido-3'-deoxy-5-methylcytidine, MedChemExpress
MedChemExpress 3'-Azido-3'-deoxy-5-methylcytidine (CS-92) is a potent xenotropic murine leukemia-related retrovirus (XMRV) inhibitor with a CC50 of 43.5 μM in MCF-7 cells. 3'-Azido-3'-deoxy-5-methylcytidine also inhibits HIV-1 reverse transcriptase with an EC50 of 0.06 μM in peripheral blood mononuclear (PBM) cells.
Brand: Medchem Express HY-111640
Description
3'-Azido-3'-deoxy-5-methylcytidine (CS-92) is a potent xenotropic murine leukemia-related retrovirus (XMRV) inhibitor with a CC50 of 43.5 μM in MCF-7 cells. 3'-Azido-3'-deoxy-5-methylcytidine also inhibits HIV-1 reverse transcriptase with an EC50 of 0.06 μM in peripheral blood mononuclear (PBM) cells.Specifications
3'-Azido-3'-deoxy-5-methylcytidine | |
Light Yellow | |
C10H14N6O4 | |
DMSO : 83.33 mg/mL (295.22 mM; Need ultrasonic) | |
282.26 | |
99.15% | |
Solid |
1282040-14-5 | |
Research | |
10 mg | |
OC[C@@H]1[C@@H](N=[N+]=[N-])[C@H]([C@H](N2C(N=C(C(C)=C2)N)=O)O1)O | |
282.26 | |
Research | |
Neuroscience-Neuromodulation |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only