Learn More
3'-Azido-3'-deoxy-5-methylcytidine, MedChemExpress
MedChemExpress 3'-Azido-3'-deoxy-5-methylcytidine (CS-92) is a potent xenotropic murine leukemia-related retrovirus (XMRV) inhibitor with a CC50 of 43.5 μM in MCF-7 cells. 3'-Azido-3'-deoxy-5-methylcytidine also inhibits HIV-1 reverse transcriptase with an EC50 of 0.06 μM in peripheral blood mononuclear (PBM) cells.
1489.66 DKK - 4458.93 DKK
Specifications
Chemical Name or Material | 3'-Azido-3'-deoxy-5-methylcytidine |
---|---|
CAS | 1282040-14-5 |
Color | Light Yellow |
Purity Grade Notes | Research |
Molecular Formula | C10H14N6O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16440817
|
Medchem Express
HY-111640 |
50 mg |
4458.93 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16424508
|
Medchem Express
HY-111640 |
10 mg |
1489.66 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3'-Azido-3'-deoxy-5-methylcytidine (CS-92) is a potent xenotropic murine leukemia-related retrovirus (XMRV) inhibitor with a CC50 of 43.5 μM in MCF-7 cells. 3'-Azido-3'-deoxy-5-methylcytidine also inhibits HIV-1 reverse transcriptase with an EC50 of 0.06 μM in peripheral blood mononuclear (PBM) cells.Specifications
3'-Azido-3'-deoxy-5-methylcytidine | |
Light Yellow | |
C10H14N6O4 | |
OC[C@@H]1[C@@H](N=[N+]=[N-])[C@H]([C@H](N2C(N=C(C(C)=C2)N)=O)O1)O | |
282.26 | |
Research | |
Neuroscience-Neuromodulation |
1282040-14-5 | |
Research | |
DMSO : 83.33 mg/mL (295.22 mM; Need ultrasonic) | |
282.26 | |
99.15% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.