Learn More
3'-Azido-3'-deoxy-5-methylcytidine, MedChemExpress
MedChemExpress 3'-Azido-3'-deoxy-5-methylcytidine (CS-92) is a potent xenotropic murine leukemia-related retrovirus (XMRV) inhibitor with a CC50 of 43.5 μM in MCF-7 cells. 3'-Azido-3'-deoxy-5-methylcytidine also inhibits HIV-1 reverse transcriptase with an EC50 of 0.06 μM in peripheral blood mononuclear (PBM) cells.
Brand:  Medchem Express HY-111640
Encompass_Preferred
 View more versions of this product
Product Code. 16424508
  • 1489.66 DKK / 10mg
Explore more special offers
Description
Description
3'-Azido-3'-deoxy-5-methylcytidine (CS-92) is a potent xenotropic murine leukemia-related retrovirus (XMRV) inhibitor with a CC50 of 43.5 μM in MCF-7 cells. 3'-Azido-3'-deoxy-5-methylcytidine also inhibits HIV-1 reverse transcriptase with an EC50 of 0.06 μM in peripheral blood mononuclear (PBM) cells.
Specifications
Specifications
3'-Azido-3'-deoxy-5-methylcytidine
Light Yellow
C10H14N6O4
DMSO : 83.33 mg/mL (295.22 mM; Need ultrasonic)
282.26
99.15%
Solid
1282040-14-5
Research
10 mg
OC[C@@H]1[C@@H](N=[N+]=[N-])[C@H]([C@H](N2C(N=C(C(C)=C2)N)=O)O1)O
282.26
Research
Neuroscience-Neuromodulation
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.