Learn More
ABT-046, MedChemExpress
MedChemExpress ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM).
Brand:  Medchem Express HY-15197
Encompass_Preferred
 View more versions of this product
Product Code. 16406368
  • 3847.22 DKK / 10mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM).
Specifications
Specifications
ABT-046
Off-White
C20H22N4O2
DMSO : 66.67 mg/mL (190.26 mM; Need ultrasonic)
350.41
98.13%
Solid
1031336-60-3
Research
10 mg
NC1=C(C2=CC=C([C@H]3CC[C@H](CC(O)=O)CC3)C=C2)C=NC4=CC=NN41
350.41
Research
Neuroscience-Neuromodulation
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
ABT-046, MedChemExpress