missing translation for 'onlineSavingsMsg'
Learn More

ABT-046, MedChemExpress

MedChemExpress ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM).

Encompass_Preferred

2286.17 DKK - 20706.45 DKK

Specifications

Chemical Name or Material ABT-046
CAS 1031336-60-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22N4O2
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16498947
Encompass_Preferred
View Documents
Medchem Express
HY-15197
5 mg
2286.17 DKK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16456666
Encompass_Preferred
View Documents
Medchem Express
HY-15197
100 mg
20706.45 DKK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16442477
Encompass_Preferred
View Documents
Medchem Express
HY-15197
50 mg
14563.00 DKK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16406368
Encompass_Preferred
View Documents
Medchem Express
HY-15197
10 mg
3847.22 DKK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM).
Specifications

Specifications

ABT-046
Off-White
C20H22N4O2
NC1=C(C2=CC=C([C@H]3CC[C@H](CC(O)=O)CC3)C=C2)C=NC4=CC=NN41
350.41
Research
Neuroscience-Neuromodulation
1031336-60-3
Research
DMSO : 66.67 mg/mL (190.26 mM; Need ultrasonic)
350.41
98.13%
Solid
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
ABT-046, MedChemExpress

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.