Learn More
ABT-046, MedChemExpress
MedChemExpress ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM).
Brand: Medchem Express HY-15197
This item is not returnable.
View return policy
Description
ABT-046 is a potent, selective, and orally efficacious acyl CoA:diacylglycerol acyltransferase 1 (DGAT-1) inhibitor (IC50= 8 nM).Specifications
ABT-046 | |
Off-White | |
C20H22N4O2 | |
DMSO : 66.67 mg/mL (190.26 mM; Need ultrasonic) | |
350.41 | |
98.13% | |
Solid |
1031336-60-3 | |
Research | |
50 mg | |
NC1=C(C2=CC=C([C@H]3CC[C@H](CC(O)=O)CC3)C=C2)C=NC4=CC=NN41 | |
350.41 | |
Research | |
Neuroscience-Neuromodulation |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 8
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
ABT-046, MedChemExpress