Learn More
Memmert™ Additional Reinforced Stainless Steel Grid (For Reinforced Chamber Option), For Ich260
Use these grids with Memmert ovens. Memmert™ Stainless Steel Reinforced Grids are designed to hold up to 60 kg.
Brand:  Memmert™ ICH260E29766
Product Code. 16316657
  • 3220.00 DKK / Each
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Description
  • Reinforced stainless steel grid
  • Max. loading 60 kg
  • From size 450 with guide bars and fixing screws and only in connection with option K1
Specifications
Specifications
60 kg
Stainless Steel
260
Model 260
Reinforced Grid
Product Title
Memmert™ Additional Reinforced Stainless Steel Grid (For Reinforced Chamber Option), For Ich260 >