missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Agar, Bacteriological Grade

Specially purified for use in preparing solid culture media for microbiological and bacteriological applications.

Brand:  MP Biomedicals™ 0215017880

 View more versions of this product

Product Code. 11456642

  • 1255.00 DKK / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • From Red alga Rhodophyceae
  • Bacteriological Grade
  • White to light yellow
  • 100 mesh powder
  • CAS: 9002-18-0
  • EC No. 232-658-1
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Red Alga Rhodophyceae
15°C to 30°C
Bacteriological
Used for preparing solid culture media for microbiological and bacteriological application
Off-white
Powder
Agar
100 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Agar, Bacteriological Grade > Quantity: 100g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.