missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Agar, Bacteriological Grade

Specially purified for use in preparing solid culture media for microbiological and bacteriological applications.

1255.00 DKK - 4295.00 DKK


Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11456642
View Documents
MP Biomedicals™
0215017880
100 g
1255.00 DKK
100g
Please to purchase this item. Need a web account? Register with us today!
11466642
View Documents
MP Biomedicals™
0215017883
250 g N/A N/A N/A
11476642
View Documents
MP Biomedicals™
0215017890
500 g
4295.00 DKK
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • From Red alga Rhodophyceae
  • Bacteriological Grade
  • White to light yellow
  • 100 mesh powder
  • CAS: 9002-18-0
  • EC No. 232-658-1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Agar, Bacteriological Grade

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.