Learn More
Thermo Scientific™ alamarBlue™ Dye
Allows three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.
Brand:  Thermo Scientific™ Y00-100
3540.07 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15559604
  • 4340.00 DKK / 100mL
Explore more special offers
Description
Description
Thermo Scientific™ alamarBlue provides unmatched sensitivity and versatility, allowing three assays to be performed with just one dye, including cell growth determination, in vitro cytotoxicity and cytokine assays.
alamarBlue, a resazurin-based product with extended incubation time, is stable for up to 5 days.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alamarBlue indicator dye
Indicator Dye
100 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number255315 Certificate TypeCertificate of Analysis Date25/04/2025 Catalog NumberY00-100
Lot Number244131 Certificate TypeCertificate of Analysis Date14/03/2025 Catalog NumberY00-100
Lot Number178495 Certificate TypeCertificate of Analysis Date02/11/2024 Catalog NumberY00-100
Lot Number155901 Certificate TypeCertificate of Analysis Date24/08/2024 Catalog NumberY00-100
Lot Number103759 Certificate TypeCertificate of Analysis Date12/05/2024 Catalog NumberY00-100
Product Title
Thermo Scientific™ alamarBlue™ Dye > 100mL