Learn More
Alfuzosin hydrochloride, MedChemExpress
MedChemExpress Alfuzosin hydrochloride is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).
Brand: Medchem Express HY-B0192A
This item is not returnable.
View return policy
Description
Alfuzosin hydrochloride is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).Specifications
Alfuzosin hydrochloride | |
White | |
C19H28ClN5O4 | |
SL 77499-10 | |
O=C(C1OCCC1)NCCCN(C2=NC(N)=C3C=C(OC)C(OC)=CC3=N2)C.Cl | |
425.91 | |
Research |
81403-68-1 | |
Research | |
50 mg | |
H2O : 50 mg/mL (117.40 mM; Need ultrasonic) ∣DMSO : 31.25 mg/mL (73.37 mM; Need ultrasonic) | |
425.91 | |
98.0% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.