Learn More
Alfuzosin hydrochloride, MedChemExpress
MedChemExpress Alfuzosin hydrochloride is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).
785.09 DKK - 2153.98 DKK
Specifications
Chemical Name or Material | Alfuzosin hydrochloride |
---|---|
CAS | 81403-68-1 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C19H28ClN5O4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16498398
|
Medchem Express
HY-B0192A |
10 mg |
785.09 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16484287
|
Medchem Express
HY-B0192A |
50 mg |
2153.98 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alfuzosin hydrochloride is an α1 adrenergic receptor antagonist used to treat benign prostatic hyperplasia (BPH).Specifications
Alfuzosin hydrochloride | |
White | |
C19H28ClN5O4 | |
H2O : 50 mg/mL (117.40 mM; Need ultrasonic) ∣DMSO : 31.25 mg/mL (73.37 mM; Need ultrasonic) | |
425.91 | |
98.0% | |
Solid |
81403-68-1 | |
Research | |
SL 77499-10 | |
O=C(C1OCCC1)NCCCN(C2=NC(N)=C3C=C(OC)C(OC)=CC3=N2)C.Cl | |
425.91 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.