missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Aluminum Evacuation Panel Safety Signs

Brand:  Brady™ 807246

 View more versions of this product

Product Code. 11760843

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Aluminum
with Up Arrow
200 mm
400 mm
Evacuation Panel Safety Sign
1 Ea.
for Indoor and Outdoor Safety Signs
120°C
Exit and emergency exit
White on Green
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Brady™ Aluminum Evacuation Panel Safety Signs > with Up Arrow; Length: 200mm; Width: 400mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.