Learn More
Astragaloside II, MedChemExpress
MedChemExpress Astragaloside II is a natural isolated from Astragalus.
Brand:  Medchem Express HY-N0433
Encompass_Preferred
 View more versions of this product
Product Code. 16441408
  • 986.40 DKK / 5mg
Explore more special offers
Description
Description
Astragaloside II is a natural isolated from Astragalus.
Specifications
Specifications
Astragaloside II
White
C43H70O15
Astrasieversianin VIII
O[C@H]1[C@H](O)[C@@H](OC(C)=O)[C@]([H])(O[C@@H]2C(C)(C)[C@@]([C@@H](O[C@]3([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)C[C@]4([H])[C@@]56CC[C@@]7(C)[C@@]4(C)C[C@H](O)[C@@]7([C@]8(C)O[C@H](C(O)(C)C)CC8)[H])([H])[C@]5(C6)CC2)OC1
827.01
Research
84676-89-1
Research
5 mg
DMSO : 100 mg/mL (120.92 mM; Need ultrasonic)
827.01
98.0%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.