Learn More
Astragaloside II, MedChemExpress
MedChemExpress Astragaloside II is a natural isolated from Astragalus.
986.40 DKK - 1781.55 DKK
Specifications
Chemical Name or Material | Astragaloside II |
---|---|
CAS | 84676-89-1 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C43H70O15 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16498978
|
Medchem Express
HY-N0433 |
10 mg |
1781.55 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16441408
|
Medchem Express
HY-N0433 |
5 mg |
986.40 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Astragaloside II is a natural isolated from Astragalus.Specifications
Astragaloside II | |
White | |
C43H70O15 | |
DMSO : 100 mg/mL (120.92 mM; Need ultrasonic) | |
827.01 | |
98.0% | |
Solid |
84676-89-1 | |
Research | |
Astrasieversianin VIII | |
O[C@H]1[C@H](O)[C@@H](OC(C)=O)[C@]([H])(O[C@@H]2C(C)(C)[C@@]([C@@H](O[C@]3([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)C[C@]4([H])[C@@]56CC[C@@]7(C)[C@@]4(C)C[C@H](O)[C@@]7([C@]8(C)O[C@H](C(O)(C)C)CC8)[H])([H])[C@]5(C6)CC2)OC1 | |
827.01 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.