Learn More
Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables
Optimize your Thermo Scientific™ Barnstead™ RO and TII water purification system with these accessories and consumables, including filters, cartridges and dispensers.
Brand: Thermo Scientific™ D502114
1033.60 DKK valid until 2025-08-24
Use promo code "24140" to get your promotional price.
Description
Optimize your water purification system with Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables. For use with Barnstead RO and TII water purification systems, these products include filters and pre-filters, cartridges, overflow cutoff float and pump interlock cable, as well as dispensers and printer.
Optimize your water purification system with Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables. For use with Barnstead RO and TII water purification systems, these products include filters and pre-filters, cartridges, overflow cutoff float and pump interlock cable, as well as dispensers and printer.
Specifications
Barnstead RO, Barnstead TII | |
Cartridge, MPS |
Promotions
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
-
Multiple Manufacturers
Focus On Clean Water
Shop now for up to 60% off Clean Water Research essentialsPromo Code: 24139Expires: 2025-08-21
New Offer Save Now