Learn More
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
11496348
|
Thermo Scientific™
AY1265X1 |
Control Panel Mounting Assembly | N/A | |||||||
10044641
|
Thermo Scientific™
D11981 |
Remote Dispenser |
8942.48 DKK
Each |
Estimated Shipment: 16-09-2025
Log in to see stock. |
||||||
10676803
|
Thermo Scientific™
D13661 |
AccuDIspense Volumetric Remote Dispenser | N/A | |||||||
10091622
|
Thermo Scientific™
D502113 |
1µm Prefilter |
1140.17 DKK
Each |
In Stock
Log in to see stock. |
||||||
10123122
|
Thermo Scientific™
D502114 |
MPS Cartridge |
1088.00 DKK
Each |
Estimated Shipment: 20-06-2025
Log in to see stock. |
||||||
10646243
|
Thermo Scientific™
D502115 |
Carbon Cartridge |
1020.93 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10429813
|
Thermo Scientific™
D502137 |
TII Filter Cartridges |
3137.32 DKK
Each |
In Stock
Log in to see stock. |
||||||
10212204
|
Thermo Scientific™
D502138 |
Start-Up Kit |
6326.80 DKK
Each |
Estimated Shipment: 02-09-2025
Log in to see stock. |
||||||
10550103
|
Thermo Scientific™
D7427 |
Reverse Osmosis Mixing Valve | N/A | |||||||
10749304
|
Thermo Scientific™
FL1265X1 |
RO Membrane |
5186.64 DKK
Each |
In Stock
Log in to see stock. |
||||||
10194803
|
Thermo Scientific™
LMX31 |
Barnstead storage reservoir ultraviolet lamp |
1753.33 DKK
Each |
In Stock
Log in to see stock. |
||||||
Description
Optimize your water purification system with Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables. For use with Barnstead RO and TII water purification systems, these products include filters and pre-filters, cartridges, overflow cutoff float and pump interlock cable, as well as dispensers and printer.
Optimize your water purification system with Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables. For use with Barnstead RO and TII water purification systems, these products include filters and pre-filters, cartridges, overflow cutoff float and pump interlock cable, as well as dispensers and printer.Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Thermo Scientific™ Barnstead™ RO and TII Accessories and Consumables