Learn More
Brady™ BBP™ 33 Series WorkHorse™ Chemical Resistant, Extreme Temperature Polypropylene Labels
BBP™ 33 Series WorkHorse™ Chemical Resistant, Extreme Temperature Polypropylene Labels are ideal for general purpose barcode applications and withstand liquid nitrogen, autoclave and other harsh environments.
1140.00 DKK - 2505.00 DKK
Specifications
Adhesive Type | Acrylic |
---|---|
Color | White |
Finish | Matte |
Material | Acrylic |
Print Technology | Thermal Transfer |
Product Code | Brand | Height (English) | Width (English) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (English) | Width (English) | Quantity | Price | Quantity & Availability | |||
17110181
|
Brady™
148677 |
0.25 in. | 0.825 in. | 1500/Pk. |
1275.00 DKK
Pack of 1500 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||
17150181
|
Brady™
148682 |
0.375 in. | 1 in. | 1500/Pk. |
1885.00 DKK
Pack of 1500 |
In Stock
Log in to see stock. |
|||
17140181
|
Brady™
148681 |
0.375 in. | 1 in. | 2000/Pk. | N/A | ||||
17170171
|
Brady™
148673 |
0.5 in. | 1 in. | 2000/Pk. |
1930.00 DKK
Pack of 2000 |
In Stock
Log in to see stock. |
|||
17130181
|
Brady™
148680 |
0.75 in. | 1 in. | 2000/Pk. |
2290.00 DKK
Pack of 2000 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||
17190171
|
Brady™
148675 |
1 in. | 1 in. | 1500/Pk. |
1890.00 DKK
Pack of 1500 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||
17150171
|
Brady™
148671 |
0.6 in. | 1.3 in. | 1500/Pk. |
1845.00 DKK
Pack of 1500 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||
17120181
|
Brady™
148679 |
0.5 in. | 1.5 in. | 1500/Pk. |
1715.00 DKK
Pack of 1500 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||
17100181
|
Brady™
148676 |
0.75 in. | 1.5 in. | 1500/Pk. |
1995.00 DKK
Pack of 1500 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||
17160171
|
Brady™
148672 |
1 in. | 1.5 in. | 1250/Pk. |
1140.00 DKK
Pack of 1250 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||
17170181
|
Brady™
148684 |
1.75 in. | 1.5 in. | 900/Pk. |
1435.00 DKK
Pack of 900 |
Estimated Shipment: 13-02-2025
Log in to see stock. |
|||
17180171
|
Brady™
148674 |
0.5 in. | 2 in. | 2000 |
2505.00 DKK
Pack of 2000 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||
17108950
|
Brady™
148678 |
0.25 in. | 2 in. | 2000/Pk. |
1930.00 DKK
Pack of 2000 |
Estimated Shipment: 15-01-2025
Log in to see stock. |
|||
17160181
|
Brady™
148683 |
1 in. | 2 in. | 1500/Pk. |
2300.00 DKK
Pack of 1500 |
In Stock
Log in to see stock. |
|||
Description
- Perfect for laboratory identification such as vials, centrifuge tubes and test tubes
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
- Service temperature of -112 to 158° F (-80 to 70° C)
Compliance
ASTM D 1000;ASTM D 2979;RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC
Specifications
Acrylic | |
Matte | |
Thermal Transfer |
White | |
Acrylic | |
BBP™33 Label Printer |