Learn More
Bel Art™ Test Tube Baskets with Lids
Perfect for dipping, transporting and storing test tubes.
349.00 DKK
Specifications
Product Type | Basket for Test Tubes |
---|---|
Autoclavable | Not Autoclavable |
Color | White |
Holds | Test Tubes |
Material | Polypropylene |
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W x H) | Price | Quantity & Availability | ||||||
10114194
|
Bel Art™
F18737-0010 |
5 x 5.18 x 4 in. (12.7 x 13.1 x 10.1cm) |
349.00 DKK
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
Description
- Polypropylene
- Can be used for transporting small lab animals
Specifications
Basket for Test Tubes | |
White | |
Polypropylene | |
Yes | |
Test Tubes |
Not Autoclavable | |
Test Tubes | |
Square | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bel Art™ Test Tube Baskets with Lids