Learn More
Kern Sohn Bench Scale ECB
Battery operation, compact, and lightweight bench scale with recessed grips on the underside.
Brand:  Kern Sohn ECB 20K-2N
Encompass_Preferred
 View more versions of this product
Product Code. 17381563
  • 1064.69 DKK / Each
Explore more special offers
Description
Description
  • High mobility: Battery operation, compact, lightweight construction, recessed grips on the underside and low weight make this balance ideal for use in several locations
  • Particularly large weighing plate
  • Very fast display: steady weight values within 2 s
  • Simple and convenient 2-key operation
Specifications
Specifications
20 kg
260 x 320 mm
10 g
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.