Learn More
Kern Sohn Bench Scale ECB
Battery operation, compact, and lightweight bench scale with recessed grips on the underside.
1064.69 DKK
Specifications
Dimensions (LxW) Platform 260 x 320 mm
View More Specs
Products 3 Description
Description
  • High mobility: Battery operation, compact, lightweight construction, recessed grips on the underside and low weight make this balance ideal for use in several locations
  • Particularly large weighing plate
  • Very fast display: steady weight values within 2 s
  • Simple and convenient 2-key operation
Specifications
Specifications
260 x 320 mm
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.