Learn More
Betulinic acid, MedChemExpress
MedChemExpress Betulinic acid is a natural pentacyclic triterpenoid, acts as a eukaryotic topoisomerase I inhibitor, with an IC50 of 5 μM, and possesses anti-HIV, anti-malarial, anti-inflammatory and anti-tumor properties.
Brand:  Medchem Express HY-10529
Encompass_Preferred
 View more versions of this product
Product Code. 16465016
  • 593.85 DKK / 100mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Betulinic acid is a natural pentacyclic triterpenoid, acts as a eukaryotic topoisomerase I inhibitor, with an IC50 of 5 μM, and possesses anti-HIV, anti-malarial, anti-inflammatory and anti-tumor properties.
Specifications
Specifications
Betulinic acid
Off-White
C30H48O3
Lupatic acid Betulic acid
[H][C@@]12CC[C@@]3(C)[C@]4(C)CC[C@]5(C(O)=O)[C@@]([C@H](C(C)=C)CC5)([H])[C@@](CC[C@@]([H])3[C@@]1(C)CC[C@H](O)C2(C)C)4[H]
456.7
Research
Cancer-programmed cell death
472-15-1
Research
100 mg
DMSO : 35.71 mg/mL (78.19 mM; Need ultrasonic)
456.7
98.0%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.