Learn More
Betulinic acid, MedChemExpress
MedChemExpress Betulinic acid is a natural pentacyclic triterpenoid, acts as a eukaryotic topoisomerase I inhibitor, with an IC50 of 5 μM, and possesses anti-HIV, anti-malarial, anti-inflammatory and anti-tumor properties.
593.85 DKK - 2083.52 DKK
Specifications
Chemical Name or Material | Betulinic acid |
---|---|
CAS | 472-15-1 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C30H48O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16481576
|
Medchem Express
HY-10529 |
500 mg |
2083.52 DKK
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16465016
|
Medchem Express
HY-10529 |
100 mg |
593.85 DKK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16420996
|
Medchem Express
HY-10529 |
200 mg |
946.13 DKK
200mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Betulinic acid is a natural pentacyclic triterpenoid, acts as a eukaryotic topoisomerase I inhibitor, with an IC50 of 5 μM, and possesses anti-HIV, anti-malarial, anti-inflammatory and anti-tumor properties.Specifications
Betulinic acid | |
Off-White | |
C30H48O3 | |
DMSO : 35.71 mg/mL (78.19 mM; Need ultrasonic) | |
456.7 | |
98.0% | |
Solid |
472-15-1 | |
Research | |
Lupatic acid Betulic acid | |
[H][C@@]12CC[C@@]3(C)[C@]4(C)CC[C@]5(C(O)=O)[C@@]([C@H](C(C)=C)CC5)([H])[C@@](CC[C@@]([H])3[C@@]1(C)CC[C@H](O)C2(C)C)4[H] | |
456.7 | |
Research | |
Cancer-programmed cell death |
Safety and Handling
H315∣H319∣H335
missing translation for 'shelfLife' : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Recommended Storage : 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.