Learn More
Sartorius™ Biohit™ Piston Assemblies
Designed for use with a wide variety of manual and electronic pipettes. Sartorius™ Biohit™ Piston Assemblies are replacement pistons for providing precise and easy pipetting operation. Available for a range of compatible pipette lines and sizes.
Brand:  Sartorius™ 721416
 View more versions of this product
Product Code. 11604572
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Controls the aspiration and dispensing operations according to selected operating mode
  • Only a quick click is required to trigger the operation of the pipette
  • If the button is kept down, the piston will stop in the lowest position until the button is released
  • Helps reduce the risk of repetitive strain injuries (RSI) that are common in pipetting
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Piston Assembly
50 to 300μL Multi Channel Pipette
Hard Tip Cone