Learn More
Sartorius™ Biohit™ Piston Assemblies
Designed for use with a wide variety of manual and electronic pipettes. Sartorius™ Biohit™ Piston Assemblies are replacement pistons for providing precise and easy pipetting operation. Available for a range of compatible pipette lines and sizes.
282.94 DKK - 358.87 DKK
Product Code | Brand | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Includes | Price | Quantity & Availability | ||||
11604572
|
Sartorius™
721416 |
50 to 300μL Multi Channel Pipette | Hard Tip Cone | N/A | |||||
11614562
|
Sartorius™
721293 |
50 to 250μL, 50 to 300μL Multi Channel Pipette | Hard Tip Cone | N/A | |||||
11654552
|
Sartorius™
721278 |
50 to 300μL Multi Channel Pipette | Tip Cone | N/A | |||||
11609132
|
Sartorius™
726154 |
300μL mLINE Multichannel, Sealpette Pro Pipette | Piston, Tip Cone and O-ring | N/A | |||||
Description
- Controls the aspiration and dispensing operations according to selected operating mode
- Only a quick click is required to trigger the operation of the pipette
- If the button is kept down, the piston will stop in the lowest position until the button is released
- Helps reduce the risk of repetitive strain injuries (RSI) that are common in pipetting
Specifications
Piston Assembly |