Learn More
Sartorius™ Biohit™ Piston Assemblies
Designed for use with a wide variety of manual and electronic pipettes. Sartorius™ Biohit™ Piston Assemblies are replacement pistons for providing precise and easy pipetting operation. Available for a range of compatible pipette lines and sizes.
Brand: Sartorius™ 721278
This item is not returnable.
View return policy
Description
- Controls the aspiration and dispensing operations according to selected operating mode
- Only a quick click is required to trigger the operation of the pipette
- If the button is kept down, the piston will stop in the lowest position until the button is released
- Helps reduce the risk of repetitive strain injuries (RSI) that are common in pipetting
Specifications
Piston Assembly | |
50 to 300μL Multi Channel Pipette | |
Tip Cone |