Learn More
Sartorius™ Biohit™ Tip Cones
Ensures a proper seal of the tip to the pipette. Sartorius™ Biohit™ Tip Cones are designed for use with a variety of mechanical and electronic single- or multi-channel pipettes. Available in several sizes for a variety of pipette models/tips.
Brand: Sartorius™ 731081
This item is not returnable.
View return policy
Description
- Compatible with Optifit tips and SafetySpace™ filter tips
- To attach a tip, simply press it onto the spring loaded tip cone for optimal sealing
- These tip holders are designed for use with single- or multi-channel Biohit™ pipettes
- Available in several sizes for a variety of mechanical and electronic units
- Made of durable thermoplastic
Specifications
Tip Cone | |
E-Line120 and Mline 100 Pipette |