Learn More
Sartorius™ Biohit™ Tip Cones
Ensures a proper seal of the tip to the pipette. Sartorius™ Biohit™ Tip Cones are designed for use with a variety of mechanical and electronic single- or multi-channel pipettes. Available in several sizes for a variety of pipette models/tips.
166.43 DKK - 492.02 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
11679122
|
Sartorius™
726151 |
300μL mLINE / Proline Plus Multichannel, Sealpette Pro Pipette | N/A | ||||||
11609302
|
Sartorius™
731081 |
E-Line120 and Mline 100 Pipette | N/A | ||||||
11624552
|
Sartorius™
721275 |
10μL Proline Electronic, Mechanical, ePET Electronic, Hamilton SofTouch Electronic Multichannel Pipette | N/A | ||||||
11604552
|
Sartorius™
721273 |
50μL Proline Mechanical Multichannel Pipette | N/A | ||||||
11624722
|
Sartorius™
726038 |
5mL mLINE / Proline Plus Single Channel Pipette, Also fits 5mL Sealpette Pro Pipette | N/A | ||||||
11644722
|
Sartorius™
726042 |
100μL mLINE Pipette | N/A | ||||||
11674442
|
Sartorius™
721012 |
2.5μL Proline Mechanical Single Channel Pipette | N/A | ||||||
Description
- Compatible with Optifit tips and SafetySpace™ filter tips
- To attach a tip, simply press it onto the spring loaded tip cone for optimal sealing
- These tip holders are designed for use with single- or multi-channel Biohit™ pipettes
- Available in several sizes for a variety of mechanical and electronic units
- Made of durable thermoplastic
Specifications
Tip Cone |