Learn More
Sartorius™ Biohit™ Tip Cones
Ensures a proper seal of the tip to the pipette. Sartorius™ Biohit™ Tip Cones are designed for use with a variety of mechanical and electronic single- or multi-channel pipettes. Available in several sizes for a variety of pipette models/tips.
166.43 DKK - 492.02 DKK
Specifications
Type Tip Cone
View More Specs
Products 7
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11679122
View Documents
Sartorius™
726151
300μL mLINE / Proline Plus Multichannel, Sealpette Pro Pipette This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11609302
View Documents
Sartorius™
731081
E-Line120 and Mline 100 Pipette This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11624552
View Documents
Sartorius™
721275
10μL Proline Electronic, Mechanical, ePET Electronic, Hamilton SofTouch Electronic Multichannel Pipette This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11604552
View Documents
Sartorius™
721273
50μL Proline Mechanical Multichannel Pipette This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11624722
View Documents
Sartorius™
726038
5mL mLINE / Proline Plus Single Channel Pipette, Also fits 5mL Sealpette Pro Pipette This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11644722
View Documents
Sartorius™
726042
100μL mLINE Pipette This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11674442
View Documents
Sartorius™
721012
2.5μL Proline Mechanical Single Channel Pipette This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Compatible with Optifit tips and SafetySpace™ filter tips
  • To attach a tip, simply press it onto the spring loaded tip cone for optimal sealing
  • These tip holders are designed for use with single- or multi-channel Biohit™ pipettes
  • Available in several sizes for a variety of mechanical and electronic units
  • Made of durable thermoplastic
Specifications
Specifications
Tip Cone