Learn More
Sartorius™ Biohit™ Tip Cones
Ensures a proper seal of the tip to the pipette. Sartorius™ Biohit™ Tip Cones are designed for use with a variety of mechanical and electronic single- or multi-channel pipettes. Available in several sizes for a variety of pipette models/tips.
Brand:  Sartorius™ 726151
 View more versions of this product
Product Code. 11679122
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Compatible with Optifit tips and SafetySpace™ filter tips
  • To attach a tip, simply press it onto the spring loaded tip cone for optimal sealing
  • These tip holders are designed for use with single- or multi-channel Biohit™ pipettes
  • Available in several sizes for a variety of mechanical and electronic units
  • Made of durable thermoplastic
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tip Cone
300μL mLINE / Proline Plus Multichannel, Sealpette Pro Pipette