Learn More
Bohlender™ BOLA™ Balls
Balls for use as splash guards or extending surfaces.
Brand: Bohlender™ H964-24
This item is not returnable.
View return policy
Description
- Made of solid PTFE, with smooth surface
- Different packing units
- For extension of surfaces such as distillation apparatus or as splash guard
Specifications
PTFE | |
30 mm | |
Smooth Surface | |
25 Pk. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bohlender™ BOLA™ Balls > Balls; Diameter (Metric): 30 mm; Material: PTFE; Quantity: 25 Pk.; Description: Smooth Surface
Spot an opportunity for improvement?Share a Content Correction