Learn More
Bohlender™ BOLA™ Balls
Balls for use as splash guards or extending surfaces.
1035.00 DKK - 3600.00 DKK
Specifications
Material PTFE
Description Smooth Surface
View More Specs
Products 6
Product Code Brand Diameter (Metric) Material Quantity Price Quantity & Availability  
Product Code Brand Diameter (Metric) Material Quantity Price Quantity & Availability  
17127424
View Documents
Bohlender™
H964-03
3 mm PTFE 100 Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17147424
View Documents
Bohlender™
H964-09
9 mm PTFE 100 Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17157424
View Documents
Bohlender™
H964-15
15 mm PTFE 50 Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17177424
View Documents
Bohlender™
H964-21
25 mm PTFE 25 Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17864084
View Documents
Bohlender™
H964-21
25 mm PTFE 25 Pk.
1700.00 DKK
Pack of 25
View alternative products
Save Up to 
»
17187424
View Documents
Bohlender™
H964-24
30 mm PTFE 25 Pk. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Made of solid PTFE, with smooth surface
  • Different packing units
  • For extension of surfaces such as distillation apparatus or as splash guard
Specifications
Specifications
PTFE
Smooth Surface
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bohlender™ BOLA™ Balls