missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Boric Acid, 4% Solution + Indicator is used in the analysis and quantitative determination of organic nitrogen.
Specifications
Specifications
pH | 4.80–5.00 |
Color | Gray to Green |
CAS | 10043-35-3, 7732-18-5 |
Concentration | 40g H3BO3 + indicator per 1L H2O |
Density | 1.00g/mL |
Odor | Odorless |
Quantity | 5 L |
Molecular Formula | BH3O3 |
Physical Form | Liquid |
MDL Number | MFCD00011337 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Safety and Handling

Product Identifier
- Boric acid 4% solution + indicator
- Danger
- Reproductive toxicity Category 1B
- H360FD-May damage fertility. May damage the unborn child.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Boric acid, Methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Boric Acid, 4% Solution + Indicator, Chem-Lab >
Spot an opportunity for improvement?Share a Content Correction