Learn More
Jaytec Glass™ Borosilicate Glass Butyrometer Tube
The Borosilicate Glass Butyrometer Tube is used to determine the fat content of dairy samples using the Gerber method.
Brand:  Jaytec Glass™ MP891
Product Code. 12398955
  • 388.00 DKK / Each
Estimated Shipment: 03-07-2025
to see stock.
Explore more special offers
Description
  • Borosilicate glass
  • Capacity and tolerances in accordance with BS ISO 488:2008 (formerly BS 696)
  • Graduated round stem
Specifications
Specifications
Borosilicate Glass
Cream Butyrometer
1%
5 g
Determination of fat content using Gerber method
0 to 70%
Product Title
Jaytec Glass™ Borosilicate Glass Butyrometer Tube > Range: 0 to 70%