Learn More
BRAND™ Borosilicate Glass Conical Ground Joint Stopper
Solid or semi-hollow with octagonal grip. BRAND™ Borosilicate Glass Conical Ground Joint Stopper is constructed from Boro 3.3. to DIN 12252 standards.
Brand: BRAND™ 144540
153.51 DKK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
- Solid or semi-hollow
- Octagonal grip
Specifications
Semi-hollow with octagonal grip | |
NS 34/35 | |
Borosilicate Glass | |
Conical |
1/Pk. | |
NS 34/35 | |
Ground Joint Stopper |