Learn More
BRAND™ Borosilicate Glass Conical Ground Joint Stopper
Solid or semi-hollow with octagonal grip. BRAND™ Borosilicate Glass Conical Ground Joint Stopper is constructed from Boro 3.3. to DIN 12252 standards.
70.55 DKK - 178.00 DKK
Specifications
Material | Borosilicate Glass |
---|---|
Product Type | Ground Joint Stopper |
Shape | Conical |
Product Code | Brand | Description | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Standard Taper Joint | Price | Quantity & Availability | |||||
10273821
|
BRAND™
144520 |
Solid with octagonal grip | NS 14/23 |
70.55 DKK
Each |
In Stock
Log in to see stock. |
|||||
10798621
|
BRAND™
144525 |
Solid with octagonal grip | NS 19/26 |
98.05 DKK
Each |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
10507461
|
BRAND™
144535 |
Semi-hollow with octagonal grip | NS 29/32 |
130.00 DKK
Each |
Estimated Shipment: 21-03-2025
Log in to see stock. |
|||||
10243681
|
BRAND™
144515 |
Solid with octagonal grip | NS 12/21 |
78.50 DKK
Each |
In Stock
Log in to see stock. |
|||||
10643012
|
BRAND™
144540 |
Semi-hollow with octagonal grip | NS 34/35 |
178.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
Description
- Borosilicate glass constrcution (Boro 3.3. to DIN 12252)
- Solid or semi-hollow
- Octagonal grip
Specifications
Borosilicate Glass | |
Ground Joint Stopper | |
Conical |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
BRAND™ Borosilicate Glass Conical Ground Joint Stopper