Learn More
BRAND™ Polypropylene Thread Adapters for Bottle-Top Dispensers
Adapt current model Dispensette™ seripettor™, Titrette™ and QuikSip™ to common bottle sizes.
Brand: BRAND™ 704328
48.11 DKK valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Description
- Polypropylene Construction
- For use with Dispensette™ seripettor™ Titrette™ and QuikSip™ Bottle-top dispensers
Specifications
Bottle Thread Adapter | |
Polypropylene | |
Dispensette™, seripettor™, Titrette™ and QuikSip™ | |
GL 33/28 |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
13Hours
9Minutes
34Seconds
43New Offer Save Up to 92%