Learn More
Fisher Chemical Buffer Solution, Colour Coded Red, pH 4, Traceable to NIST, 2.5L
Brand: Fisher Chemical J/2826/17
Buffer Solution, Colour Coded Red, pH 4, Traceable to NIST, 2.5L
Description

Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
47Hours
5Minutes
13Seconds
59 -
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
Days
88Hours
5Minutes
13Seconds
59New Offer Save Up to 59%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.