Learn More
Butenafine Hydrochloride, MedChemExpress
MedChemExpress Butenafine Hydrochloride (KP363 Hydrochloride) is a synthetic benzylamine antifungal, works by inhibiting the synthesis of sterols by inhibiting squalene epoxidase.
Brand:  Medchem Express HY-17396
Encompass_Preferred
 View more versions of this product
Product Code. 16449827
  • 3293.30 DKK / 5g
Explore more special offers
This item is not returnable. View return policy
Description
Description
Butenafine Hydrochloride (KP363 Hydrochloride) is a synthetic benzylamine antifungal, works by inhibiting the synthesis of sterols by inhibiting squalene epoxidase.
Specifications
Specifications
Butenafine Hydrochloride
101827-46-7
5 g
C23H28ClN
KP363 Hydrochloride
CN(CC1=CC=C(C(C)(C)C)C=C1)CC2=C3C=CC=CC3=CC=C2.[H]Cl
353.93
Research
Solid
White
Research
COVID-19-immunoregulation
DMSO : 25 mg/mL (70.64 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.83 mM; Need ultrasonic)
353.93
99.96%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Butenafine Hydrochloride, MedChemExpress