Learn More
Butenafine Hydrochloride, MedChemExpress
MedChemExpress Butenafine Hydrochloride (KP363 Hydrochloride) is a synthetic benzylamine antifungal, works by inhibiting the synthesis of sterols by inhibiting squalene epoxidase.

684.84 DKK - 3293.30 DKK
Specifications
Chemical Name or Material | Butenafine Hydrochloride |
---|---|
Physical Form | Solid |
CAS | 101827-46-7 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16481669
![]() |
Medchem Express
HY-17396 |
1 g |
1107.84 DKK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16451846
![]() |
Medchem Express
HY-17396 |
500 mg |
684.84 DKK
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16449827
![]() |
Medchem Express
HY-17396 |
5 g |
3293.30 DKK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Butenafine Hydrochloride (KP363 Hydrochloride) is a synthetic benzylamine antifungal, works by inhibiting the synthesis of sterols by inhibiting squalene epoxidase.Specifications
Butenafine Hydrochloride | |
101827-46-7 | |
Research | |
COVID-19-immunoregulation | |
DMSO : 25 mg/mL (70.64 mM; Need ultrasonic) ∣H2O : 1 mg/mL (2.83 mM; Need ultrasonic) | |
353.93 | |
99.96% |
Solid | |
White | |
C23H28ClN | |
KP363 Hydrochloride | |
CN(CC1=CC=C(C(C)(C)C)C=C1)CC2=C3C=CC=CC3=CC=C2.[H]Cl | |
353.93 | |
Research |
Safety and Handling
H302∣H315∣H319∣H335
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.