Learn More
Azpack™ Carbon Chrome Steel Spheres
Demonstrates Archimedes' Principle and other properties of physics and math. Azpack™ Carbon Chrome Steel Spheres are used mainly for educational purposes.
Brand:  Azpack™ 12403090
 View more versions of this product
Product Code. 12403090
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Manufactured from carbon chrome steel.
  • Available in a variety of sizes
  • Smooth, polished chrome surface
  • Round-shaped balls
Specifications
Specifications
Metal Ball
Carbon Chrome Steel
Dia: 12mm
12 mm
Product Title
Azpack™ Carbon Chrome Steel Spheres > 12mm