Learn More
CAY10603, MedChemExpress
MedChemExpress CAY10603 (BML-281) is a potent and selective HDAC6 inhibitor, with an IC50 of 2 pM; CAY10603 (BML-281) also inhibits HDAC1, HDAC2, HDAC3, HDAC8, HDAC10, with IC50s of 271, 252, 0.42, 6851, 90.7 nM.
Brand:  Medchem Express HY-18613
Encompass_Preferred
 View more versions of this product
Product Code. 16441739
  • 745.27 DKK / 1mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
CAY10603 (BML-281) is a potent and selective HDAC6 inhibitor, with an IC50 of 2 pM
Specifications
Specifications
CAY10603
1045792-66-2
1 mg
C22H30N4O6
BML-281
O=C(OC(C)(C)C)NC1=CC=C(C2=CC(C(NCCCCCCC(NO)=O)=O)=NO2)C=C1
446.5
Research
Solid
Light Nude
Research
Cancer-programmed cell death
DMSO : ≥ 50 mg/mL (111.98 mM)
446.5
95.39%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
CAY10603, MedChemExpress