Learn More
CAY10603, MedChemExpress
MedChemExpress CAY10603 (BML-281) is a potent and selective HDAC6 inhibitor, with an IC50 of 2 pM; CAY10603 (BML-281) also inhibits HDAC1, HDAC2, HDAC3, HDAC8, HDAC10, with IC50s of 271, 252, 0.42, 6851, 90.7 nM.

745.27 DKK - 10353.23 DKK
Specifications
Chemical Name or Material | CAY10603 |
---|---|
Physical Form | Solid |
CAS | 1045792-66-2 |
Color | Light Nude |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16473557
![]() |
Medchem Express
HY-18613 |
50 mg |
6868.58 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16457538
![]() |
Medchem Express
HY-18613 |
10 mg |
2245.89 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16453606
![]() |
Medchem Express
HY-18613 |
25 mg |
4370.91 DKK
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16447486
![]() |
Medchem Express
HY-18613 |
100 mg |
10353.23 DKK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16441739
![]() |
Medchem Express
HY-18613 |
1 mg |
745.27 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16440018
![]() |
Medchem Express
HY-18613 |
5 mg |
1369.68 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CAY10603 (BML-281) is a potent and selective HDAC6 inhibitor, with an IC50 of 2 pMSpecifications
CAY10603 | |
1045792-66-2 | |
Research | |
Cancer-programmed cell death | |
DMSO : ≥ 50 mg/mL (111.98 mM) | |
446.5 | |
95.39% |
Solid | |
Light Nude | |
C22H30N4O6 | |
BML-281 | |
O=C(OC(C)(C)C)NC1=CC=C(C2=CC(C(NCCCCCCC(NO)=O)=O)=NO2)C=C1 | |
446.5 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.