Learn More
CAY10603, MedChemExpress
MedChemExpress CAY10603 (BML-281) is a potent and selective HDAC6 inhibitor, with an IC50 of 2 pM; CAY10603 (BML-281) also inhibits HDAC1, HDAC2, HDAC3, HDAC8, HDAC10, with IC50s of 271, 252, 0.42, 6851, 90.7 nM.
Brand: Medchem Express HY-18613
Description
CAY10603 (BML-281) is a potent and selective HDAC6 inhibitor, with an IC50 of 2 pMSpecifications
CAY10603 | |
1045792-66-2 | |
100 mg | |
C22H30N4O6 | |
BML-281 | |
O=C(OC(C)(C)C)NC1=CC=C(C2=CC(C(NCCCCCCC(NO)=O)=O)=NO2)C=C1 | |
446.5 | |
Research |
Solid | |
Light Nude | |
Research | |
Cancer-programmed cell death | |
DMSO : ≥ 50 mg/mL (111.98 mM) | |
446.5 | |
95.39% |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only