Learn More
WTW™ Chemical Oxygen Demand (COD) Test Kits
Get accurate Chemical Oxygen Demand (COD) test results with the Chlorine dioxide reagent test kit.
Brand: WTW™ 251991
716.25 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
- Variety of instrumentation for measuring Chemical Oxygen Demand (COD)
- Safe determinations of COD in water
- Chlorine dioxide (ClO<sub>2</sub>) reagent test kit
- For waste water, production control, ground water, surface water
- For PhotoLab® S6, PhotoLab® S12, PhotoLab® Spectral, spectroFlex photometers
Specifications
For waste water, production control, ground water, surface water | |
Certificates | |
20 to 1500 mg/L |
Photometers | |
90 | |
Zinc (Zn) reagent test kit |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- COD2 TC (MR)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Germ cell mutagenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 1
- Carcinogenicity Category 1B
- Specific target organ toxicity after repeated exposure Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H373-May cause damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH208-Contains potassium dichromate. May produce an allergic reaction.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : sulphuric acid, mercury sulphate, disilver sulphate, potassium dichromate