missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ C18 ec Glass Column

Brand:  Macherey-Nagel™ 730015G

 View more versions of this product

Product Code. 10104901

  • 1935.00 DKK / Pack of 30

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Silica
6 mL
1000 mg
45 μm
Solid Phase Extraction Column
Hydrophobic
500 m2/g
14%
SPE
Solid Phase Extraction
Chromabond C18 ec
Yes
60 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Macherey-Nagel™ Chromabond™ C18 ec Glass Column > Column Capacity: 6mL; Adsorbent Weight: 1000mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.